Learn More
Selleck Chemical LLC Mivacurium chloride
SDP
Supplier:  Selleck Chemical LLC S56735MG
Encompass_Preferred
Mivacurium chloride is a rapid non-depolarising neuromuscular blocking agent of short duration used therefore as anesthetic to provide skeletal muscle relaxation in minor surgical operations in emergency surgical procedures of short to intermediate duration and during intubation of the trachea
Catalog No. 50-000-03362