Learn More
Medchemexpress LLC HY-100015 25mg , Mivebresib CAS:1445993-26-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10001525MG
Encompass_Preferred
HY-100015 25mg Mivebresib CAS: 1445993-26-9 Mivebresib (ABBV-075) is a potent and orally active bromodomain and extraterminal domain (BET) bromodomain inhibitor. Mivebresib binds to BRD4 with a Ki of 1.5 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0342