Learn More
USA Scientific Inc Mixer HC ISO Rack
Supplier:  USA Scientific Inc 80120021
Encompass
Mixer HC ISO Rack for 24 x 1.5 or 2.0 mL microcentrifuge tubes, polycarbonate. Includes lid.
Catalog No. 50-274-5063