Learn More
Yamato Scientific America, Inc. Yamato Flask Mixer, Max 1000 rpm, Equipped with digital indicator, 220V.
Supplier:  Yamato Scientific America, Inc. LM210
Encompass_Preferred
Compact design laboratory flask stirrer featuring integrated drive and stirring seal allowing direct installation of flask and stirring in a vacuum and sealed state. Equipped with digital indicator. Wide range rotation speed of 50-1000 rpm. Maintenance free and superior DC brushless motor. Belt drive transmission minimizes noise and vibration. Capable of vacuuming up to 399.9Pa without impairing rotation efficiency. Directly attachable to a three-neck flask 24/40, 29/42 optional. Variety of stirring shafts and blades available to handle small to large volume samples. Fluorine rubber seal as standard for shaft seal, superior chemical resistant Teflonrubber seal available as option. At the flask joint, FKM o-ring is used as standard, superior chemical resistant Kalrez o-ring available as option. Equipped with 24/40 rotary joint, 29/42 optional. Operating speed range 50-1000 rpm.
Catalog No. 50-189-8187