Learn More
Medchemexpress LLC Mizolastine | 108612-45-9 | 99.82% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB0164500MG
Encompass_Preferred
Mizolastine is an orally active, high affinity, and specific peripheral histamine H1 receptor antagonist (second-generation antihistamine). It effectively inhibits mRNA expression of VEGF165, VEGF120, TNF-α, and KC. Mizolastine can be used in studies of allergic rhinitis and chronic idiopathic urticaria.
  • Acts as a high-affinity peripheral histamine H1 receptor antagonist
  • Inhibits mRNA expression of VEGF165, VEGF120, TNF-α, and KC
  • Useful in studies of allergic rhinitis
  • Applicable in research on chronic idiopathic urticaria
Catalog No. 50-004-06462