Learn More
Selleck Chemical LLC Mizoribine 10mg 50924-49-7 Bredinin, NSC 289637
SDP
Supplier:  Selleck Chemical LLC S138410MG
Encompass_Preferred
Mizoribine (Bredinin, NSC 289637) is an imidazole nucleoside and an immunosuppressant that inhibits HCV RNA replication with IC50 of ~ 100 ?M for anti-HCV activity. Mizoribine is a selective inhibitor of inosine-5'-monophosphate dehydrogenase (IMPDH) and guanosine monophosphate synthetase. Mizoribine also inhibits SARS-CoV. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-801-5