Learn More
Medchemexpress LLC Mizoribine (Standard) | 50924-49-7 | 99.9% | 259.22 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY17470R5MG
Encompass_Preferred
Mizoribine (Standard) is the analytical standard of Mizoribine, intended for research and analytical applications. This imidazole nucleoside inhibits HCV RNA replication with an IC50 of approximately 100 μM for anti-HCV activity. It acts as an immunosuppressant and an IMPDH inhibitor, also inhibiting the replication of SARS-CoV with IC50s of 3.5 μg/mL for SARS-CoV Frankfurt-1 and 16 μg/mL for SARS-CoV HKU39849.
  • Analytical standard for research and analytical applications
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
  • Inhibits HCV RNA replication with an IC50 of approximately 100 μM
  • Acts as an immunosuppressant
  • Inhibits replication of SARS-CoV with IC50s of 3.5 μg/mL and 16 μg/mL
  • Solid, white to off-white appearance
Catalog No. 50-003-87051