Learn More
Cayman Chemical MJ33lIthIum salt 10mg
SDP
Supplier:  Cayman Chemical 9000184410 MG
Encompass_Preferred
A selective, reversible inhibitor of the acidic, calcium-independent (ai)PLA2 activity of Prdx6, with optimal inhibition achieved at an MJ33 concentration of 3 mol%; blocks the degradation of DPPC in both whole lung and isolated alveolar type II epithelial cells
Catalog No. 50-289-4430