Learn More
Medchemexpress LLC MK-0493 (hydrochloride) | 455957-71-8 | 98.8% | 582.55 | C30H39Cl2F2N3O2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY118930A10MM 1ML
Encompass_Preferred
MK-0493 hydrochloride is a potent, orally active, and selective agonist of the melanocortin receptor 4 (MC4R), provided as a 10 mM solution in DMSO (1 mL) for preclinical research. The compound is characterized with confirmed purity and molecular data to support reproducible metabolic and neuronal signaling studies.
  • Potent and selective MC4R agonist for metabolic research.
  • Ready-to-use 10 mM solution in DMSO (1 mL) for immediate experiments.
  • High purity suitable for preclinical work.
  • Characterized molecular weight and formula provided for reproducibility.
  • Also available as solid quantities for dosing flexibility.
Catalog No. 50-003-07909