Learn More
Cayman Chemical MK-212hydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 3608910 MG
Encompass_Preferred
A 5-HT2C receptor agonist (EC50 = 0.028 µM in a calcium mobilization assay); selective for 5-HT2C over 5-HT2A receptors (EC50 = 0.42 µM); reduces food intake in fasted rats (ED50 = 5 mg/kg); reduces cocaine-primed reinstatement of cocaine-seeking behavior in rats at 0.32, 0.56, or 1 mg/kg
Catalog No. 50-291-1755