Learn More
Selleck Chemical LLC MK-2206 2HCl S1078-100mg
SDP
Supplier:  Selleck Chemical LLC S1078100MG
Encompass_Preferred
MK-2206 2HCl is a highly selective inhibitor of Akt1/2/3 with IC50 of 8 nM/12 nM/65 nM in cell-free assays respectively no inhibitory activities against 250 other protein kinases observed MK-2206 2HCl induces autophagy and apoptosis in cancer cells Phase 2
Catalog No. 50-310-8462