Learn More
Medchemexpress LLC MK-2206 dihydrochloride | 1032350-13-2 | MFCD14584463 | 99.9% | 480.4 g/mol | C25H23Cl2N5O | 25 MG
SDP
Supplier:  Medchemexpress LLC HY1035825MG
Encompass_Preferred
MK-2206 dihydrochloride is an orally active, brain-penetrant allosteric inhibitor of Akt used in preclinical research to modulate PI3K/Akt signaling. It selectively inhibits Akt1, Akt2, and Akt3 with low-nanomolar potency and has been reported to induce autophagy in cellular models.
  • Allosteric Akt inhibitor with low-nanomolar IC50s for Akt1, Akt2, and Akt3.
  • Induces autophagy in cell-based assays.
  • Orally active and brain penetrant, suitable for in vivo studies.
  • Supplied as a solid powder, soluble in DMSO for solution preparation.
  • High purity (>99%), supporting reproducible experimental results.
  • Suitable for in vitro and preclinical in vivo research applications.
Catalog No. 50-002-45019