Learn More
AdipoGen MK-2206 dihydrochloride, 1032350-13-2, MFCD14584463, 1 mg
SDP
Supplier:  AdipoGen SYN1162M001
Encompass_Preferred
MF: C25H21N5O . 2HCl, MW: 480.397, ≥95%, Appearance: Solid. MK-2206 is a highly selective inhibitor of Akt1, Akt2 and Akt3 with IC(50) of 8nM, 12nM and 65nM, respectively.
Catalog No. 50-101-5869