Learn More
Cayman Chemical MK-2206hydrochlorIde 5mg
SDP
Supplier:  Cayman Chemical 115935 MG
Encompass_Preferred
An orally active, allosteric Akt inhibitor (IC50s = 5, 12, and 65 nM for Akt1, 2, and 3, respectively); inhibits cell proliferation of human cancer cell lines synergistically in combination with other topoisomerase inhibitors, antimetabolites, antimicrotubule agents, DNA cross-linkers or growth factor inhibitors
Catalog No. 50-295-1783