Learn More
TARGETMOL CHEMICALS INC MK-2295 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T334295MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. MK-2295 is a potent TRPV1 antagonist. MK-2295 can be used in studies about the treatment of chronic pain. purity: 98%
Catalog No. 50-258-0307