Learn More
Selleck Chemical LLC MK-28
SDP
Supplier:  Selleck Chemical LLC S69521G
Encompass_Preferred
MK-28 is a potent and selective activator of PERK MK-28 exhibits an IC50 3-fold higher than CCT020312 in rescuing mutant huntingtin protein (mHtt)-expressing striatal neurons from apoptosis MK-28 exhibits remarkable pharmacokinetic properties and high BBB penetration in mice
Catalog No. 50-000-02241