Learn More
TARGETMOL CHEMICALS INC MK-3903 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T518710MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MK-3903 is a potent and selective AMPK activator (EC50 8 nM). purity: 98%
Catalog No. 50-257-9797