Learn More
Medchemexpress LLC Mk-571 | 115104-28-4 | 98.85% | 515.09 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1998950MG
Encompass_Preferred
MK-571 is an orally active, potent, and selective competitive leukotriene D4 (LTD4) receptor antagonist. It demonstrates Ki values of 0.22 nM in guinea pig lung and 2.1 nM in human lung membranes. This compound also functions as an MRP4 and ABCC1 (MRP1) inhibitor, effectively inhibiting constitutive and antigen-stimulated S1P (sphingosine-1-phosphate) release.
  • Orally active
  • Potent and selective competitive leukotriene D4 (LTD4) receptor antagonist
  • Acts as an MRP4 and ABCC1 (MRP1) inhibitor
  • Inhibits constitutive and antigen-stimulated S1P release
  • Recommended storage at 4°C, protected from light
Catalog No. 50-004-04527