Learn More
Medchemexpress LLC MK-571 (L-660711) | 115104-28-4 | 98.85% | 515.09 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY19989100MG
Encompass_Preferred
MK-571 (L-660711) is an orally active, potent and selective competitive leukotriene D4 (LTD4) receptor antagonist, with Ki values of 0.22 and 2.1 nM in guinea pig and human lung membranes, respectively. It is also a MRP4 and ABCC1 (MRP1) inhibitor, and it inhibits constitutive and antigen-stimulated S1P (sphingosine-1-phosphate) release.
  • Potent and selective LTD4 receptor antagonist.
  • Inhibits MRP4 and ABCC1 (MRP1).
  • Suppresses constitutive and antigen-stimulated S1P secretion.
  • Blocks LTD4- and Ascaris-induced bronchoconstriction.
  • Reverses hypoxic pulmonary hypertension (PH).
Catalog No. 50-004-03930