Learn More
Selleck Chemical LLC (+)-MK 801 maleate 10mg 77086-22-7
SDP
Supplier:  Selleck Chemical LLC S287610MG
Encompass_Preferred
(+)-MK-801 is a potent, selective and non-competitive NMDA receptor antagonist with Kd of 37.2 nM in rat brain membranes. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3123