Learn More
Abcam (-)-MK 801 maleate (mM/ml), NMDA antagonist, 20MM_ML
Supplier:  Abcam AB14666820MM_ML
Encompass_Preferred
MW 337.4 Da, Purity >99%. (-)-MK-801 maleate is a less active enantiomer of the NMDA receptor antagonist (+)-MK-801.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4536