Learn More
Medchemexpress LLC HY-13070 2mg Medchemexpress, MK-8245 CAS:1030612-90-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY130702MG
Encompass_Preferred
Medchemexpress, HY-13070 2mg MK-8245 CAS:1030612-90-8 MK-8245 is a potent, liver-targeted stearoyl-CoA desaturase (SCD) inhibitor, with IC50s of 1 nM for human SCD1 and 3 nM for both rat SCD1 and mouse SCD1, with antidiabetic and antidyslipidemic efficacy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1248