Learn More
TARGETMOL CHEMICALS INC MK-8617 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T410625MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MK-8617 is an orally available HIF PHD1 3 pan-inhibitor inhibiting PHD1/2/3 (IC50 1.0/1.0/14 nM). purity: 99%
Catalog No. 50-257-9964