Learn More
Apexbio Technology LLC MK-8745, 10mg. Cas: 885325-71-3 MFCD: MFCD28009367. Aurora A inhibitor,potent and selective
SDP
Supplier:  Apexbio Technology LLC A880710
Encompass_Preferred
MK-8745 is a novel and selective inhibitor of Aurora A with IC50 value of 0.6 nM [1].Aurora A kinase is a member of mitotic serine/threonine kinases and plays an important role in cellular division by controlling chromatid segregation. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5193