Learn More
Enzo Life Sciences MK-886 (25mg). CAS: 118414-82-7
SDP
Supplier:  Enzo Life Sciences BMLEI2660025
Encompass_Preferred
Inhibitor of PPARα activation. Inhibits 5-lipoxygenase-activating protein (FLAP) and leukotriene biosynthesis. Binds to FLAP with high affinity and prevents the activation of 5-lipoxygenase. IC50’s for leukotriene biosynthesis: 2.5 nM in intact leukocytes; 1.1 µM in whole blood. Induces apoptosis through the mitochondrial pathway. Alternative name: L-663,536, 3-[1-(p-Chlorobenzyl)-5-(isopropyl)-3-t-butylthioindol-2-yl]-2,2-dimethylpropanoic acid, Na. Purity: ≥98% (HPLC). Solubility: Soluble in 100% ethanol (25mg/ml) or DMSO (25mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0646