Learn More
Cayman Chemical mK1trIfluoroacetate salt 10mg
SDP
Supplier:  Cayman Chemical 3153610 MG
Encompass_Preferred
An FPR2 agonist; induces calcium flux in CHO cells expressing human FPR2 and Gα16 (EC50 = α16 alone or in combination with FPR1 (EC50s = >10,000 nM for both); induces chemotaxis of isolated human peripheral blood monocytes and neutrophils in a concentration-dependent manner; increases production of IL-1β and IL-6 in isolated human monocytes at 10 µM
Catalog No. 50-299-1599