Learn More
Selleck Chemical LLC MK6-83
SDP
Supplier:  Selleck Chemical LLC S694725MG
Encompass_Preferred
MK6-83 is a novel Transient receptor potential melastatin member 1 (TRPML1) activator that may be used as a potential pharmacological treatment option for a specific subgroup of MLIV patients
Catalog No. 50-000-02496