Learn More
TARGETMOL CHEMICALS INC MK8722 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1609910MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. MK8722 is an effective and systemic activator of pan-AMPK. Purity 99.88%
Catalog No. 50-287-7714