Learn More
Medchemexpress LLC MKA031 | 2951012-00-1 | 98.7% | 403.46 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1493085MG
Encompass_Preferred
MKA031 (compound 6y) is a non-competitive MIF inhibitor with an IC50 value of 1.7 μM. It interferes with MIF/AIF interaction, MIF nuclear translocation, and N-methyl-N'-nitro-N-nitrosoguanidine (MNNG)-induced dependent cell death. MKA031 can be utilized in the study of chronic hepatitis C virus infection.
  • Interferes with MIF/AIF interaction
  • Blocks MIF nuclear translocation
  • Prevents N-methyl-N'-nitro-N-nitrosoguanidine (MNNG)-induced dependent cell death
  • Protects genomic DNA by blocking the recruitment of MIF to AIF
Catalog No. 50-003-60881