Learn More
Selleck Chemical LLC MKC8866 2mg
SDP
Supplier:  Selleck Chemical LLC S88752MG
Encompass_Preferred
MKC8866 (IRE1-IN-8866), a salicylaldehyde analog, is a specific IRE1? RNase inhibitor with IC50 of 0.29??M for human IRE1? in vitro. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3413