Learn More
Selleck Chemical LLC MKC8866 S8875-25MG
SDP
Supplier:  Selleck Chemical LLC S887525MG
Encompass_Preferred
MKC8866 (IRE1-IN-8866) a salicylaldehyde analog is a specific IRE1 RNase inhibitor with IC50 of 0 29 M for human IRE1 in vitro
Catalog No. 50-000-24817