Learn More
TARGETMOL CHEMICALS INC MKT-077 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T515210MG
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MKT-077 (FJ-776) is a cationic rhodacyanine dye that demonstrates antiproliferative activity against cancer cell lines (EC50s 1.4-2.2 uM in vitro) through its ability to inhibit members of the Hsp70 family of molecular chaperones. purity: 98%
Catalog No. 50-257-9803