Learn More
Medchemexpress LLC ML-00253764 hydrochloride | 1706524-94-8 | 99.29% | 413.71 | 10 MM * 1 ML
Supplier: Medchemexpress LLC HY11012310MM/1ML
ML-00253764 hydrochloride is a brain penetrant nonpeptidic melanocortin receptor 4 (MC4R) antagonist with a Ki and IC50 of 0.16 μM and 0.103 μM, respectively. It can cross the blood-brain barrier.
- Acts as a melanocortin receptor 4 (MC4R) antagonist.
- Crosses the blood-brain barrier.
- Decreases cAMP production in MC4R-expressing HEK293 cell membranes.
- Demonstrates in vivo protection against tumor-induced body weight loss in mouse models.
- Off-white to light yellow solid appearance.