Learn More
TARGETMOL CHEMICALS INC ML-098 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T461925MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ML-098 (CID-7345532) is an activator of the GTP-binding protein Rab7 (EC50 77.6 nM). purity: 99%
Catalog No. 50-257-9861