Learn More
TARGETMOL CHEMICALS INC ML 10302 HYDROCHLORIDE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T22992
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. ML 10302 hydrochloride is a specific agonist 5-HT4 and binds 5-HT receptors with Kis of 1.07 nM and 782 nM for 5-HT4 and 5-HT3. purity: 100%
Catalog No. 50-258-0903