Learn More
Medchemexpress LLC HY-101844 5mg Medchemexpress, ML-18 CAS:1422269-30-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1018445MG
Encompass_Preferred
Medchemexpress, HY-101844 5mg ML-18 CAS:1422269-30-4 ML-18 is a non-peptide bombesin receptor subtype-3 (BRS-3) antagonist with an IC50 of 4.8 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7885