Learn More
Medchemexpress LLC ML-210 1g
SDP
Supplier:  Medchemexpress LLC HY1000031G
Encompass_Preferred
ML-210 is a selective and covalent glutathione peroxidase 4 (GPX4) inhibitor with an EC50 of 30 nM ML-210 binds the GPX4 selenocysteine residue ML-210 has anti-cancer activity1]2]
Catalog No. 50-001-68934