Learn More
Apexbio Technology LLC ML 228 1357171-62-0 50mg
SDP
Supplier:  Apexbio Technology LLC A450850
Encompass_Preferred
ML 228 (CAS 1357171-62-0) is a small molecule that stimulates the hypoxia-inducible factor (HIF) signaling pathway by chelating iron through a mechanism independent of prolyl hydroxylase domain (PHD) enzymes It enhances HIF pathway activity with EC sub 50 /sub values of 1 23 M in HRE gene reporter assays and 1 4 M for HIF-1 nuclear translocation At 10 M ML 228 also demonstrates greater than 80% inhibition of the human A sub 3 /sub adenosine receptor dopamine transporter opioid receptor hERG channel 5-HT sub 2B /sub receptor and rat sodium channel site 2 ML 228 is valuable for studies investigating HIF pathway regulation hypoxia response and off-target pharmacological profiling in cellular models
Catalog No. 50-000-94355