Learn More
Apexbio Technology LLC ML 281 200mg
SDP
Supplier:  Apexbio Technology LLC B5750200
Encompass_Preferred
ML 281 (CAS 1404437-62-2) is a small-molecule inhibitor targeting serine/threonine kinase STK33 It is designed to block the phosphorylation activity of STK33 thereby interfering with downstream signaling pathways regulated by STK33-mediated phosphorylation events ML 281 exerts its biological activity primarily through selective inhibition of STK33 kinase activity In biochemical assays ML 281 demonstrates potent inhibitory effects with an IC50 value of approximately 14 nM against STK33 Based on these pharmacological properties ML 281 holds research potential in investigating the biological role of STK33 in tumor cell survival cellular proliferation and oncogenic signal transduction pathways
Catalog No. 50-000-92625