Learn More
Enzo Life Sciences ML-7. HCl (10mg). CAS: 109376-83-2
SDP
Supplier:  Enzo Life Sciences BMLEI1970010
Encompass_Preferred
Potent and selective inhibitor of MLC kinase (Ki=0.3 µM). It inhibts also smooth-muscle myosin light chain kinase, protein kinase C (PKC) and cAMP-dependent protein kinase (PKA). Alternative name: 1-(5-Iodonaphthalene-1-sulfonyl)-1H-hexahydro-1,4-diazepine . HCl. Purity: ≥99%. Solubility: Soluble in DMSO (10mg/ml), ethanol/water (1:1, 10mg/ml), 100% ethanol (warm) and methanol. Long Term Storage: -20°C.
Catalog No. 50-201-0604