Learn More
Medchemexpress LLC ML-7 hydrochloride | 110448-33-4 | C15H18ClIN2O2S | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY15417
Encompass_Preferred
ML-7 hydrochloride is a naphthalene sulphonamide derivative that potently inhibits Myosin Light Chain Kinase (MLCK) with an IC₅₀ of 300 nM, and also inhibits YAP/TAZ. This product is intended for research use only.
  • Potently inhibits Myosin Light Chain Kinase (MLCK) with an IC₅₀ of 300 nM.
  • Inhibits YAP/TAZ.
  • Inhibits rabbit portal vein α1-adrenoceptor NSCC with an IC₅₀ of 0.8 μM.
  • Attenuates Dexmedetomidine (DMT)-induced contraction.
  • Reduces intracerebral Evans Blue extravasation, suggesting a reduction in brain edema.
  • Reduces brain water content and improves neurological function after controlled cortical impact (CCI).
  • Significantly reduces the protein level of phosphorylated MLC2 and the phosphorylation ratio of MLC2 in BMECs.
  • Directly inhibits the expression of ZO-1.
Catalog No. 50-003-75258