Learn More
Medchemexpress LLC ML-7 hydrochloride | 110448-33-4 | C15H18ClIN2O2S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY154175MG
Encompass_Preferred
ML-7 hydrochloride is a naphthalene sulphonamide derivative that potently inhibits Myosin Light Chain Kinase (MLCK) with an IC50 of 300 nM. It also acts as an inhibitor of YAP/TAZ. In vitro studies indicate its ability to inhibit rabbit portal vein α1-adrenoceptor NSCC and attenuate Dexmedetomidine (DMT)-induced contraction. In vivo, ML-7 hydrochloride has been shown to reduce brain edema formation after traumatic brain injury in mice.
  • Inhibits Myosin Light Chain Kinase (MLCK)
  • Acts as an inhibitor of YAP/TAZ
  • Inhibits rabbit portal vein α1-adrenoceptor NSCC
  • Attenuates Dexmedetomidine (DMT)-induced contraction
  • Reduces brain edema formation after traumatic brain injury in mice
Catalog No. 50-003-76842