Learn More
Medchemexpress LLC HY-108702 5mg Medchemexpress, ML-792 CAS:1644342-14-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1087025MG
Encompass_Preferred
Medchemexpress, HY-108702 5mg ML-792 CAS:1644342-14-2 ML-792 is a specific small ubiquitin-like modifier (SUMO)-activating enzyme (SAE) inhibitor. ML-792 is a potent and selective inhibitor of SAE/SUMO1 and SAE/SUMO2 in enzymatic assays (IC50s of 3 and 11 nM, respectively) compared with NAE/NEDD8 and UAE/ubiquitin (IC50s of 32 μM and >100 μM, respectively). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8614