Learn More
TARGETMOL CHEMICALS INC ML-SI3 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T922610MG
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. ML-SI3 is an inhibitor of the TRPML family of calcium channels that selectively inhibits TRPML1, TRPML2, and TRPML3. ML-SI3 can be used to study neurodegenerative and cardiovascular diseases. Purity 99.94%
Catalog No. 50-287-7034