Learn More
Abcam ML130, NOD1 inhibitor, 5MG
Supplier:  Abcam AB1421775MG
Encompass_Preferred
MW 287.34 Da, Purity >97%. Potent, selective NOD1 inhibitor (IC₅₀ = 560 nM). Inhibits NOD1-induced NF-κB activation in vitro. Cell-permeable.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6290