Learn More
Medchemexpress LLC ML133 hydrochloride | 1222781-70-5 | 313.82 g/mol | C19H20ClNO | 1 ML
SDP
Supplier:  Medchemexpress LLC HY100230A10MM 1ML
Encompass_Preferred
ML133 hydrochloride is a selective inhibitor of Kir2 family inward-rectifier potassium channels used in ion channel research and electrophysiology. It displays pH-dependent potency with reported IC50 values of 1.8 μM at pH 7.4 and 290 nM at pH 8.5. Supplied as a hydrochloride salt and commonly provided as a 10 mM solution in DMSO for laboratory research use only.
  • Selective Kir2 family inhibitor suitable for electrophysiology studies.
  • pH-dependent potency with IC50 values at physiological and alkaline pH.
  • Convenient 10 mM solution in DMSO for immediate experimental use.
  • Hydrochloride salt form for improved solubility in polar solvents.
  • Provided for research use only; not for human or veterinary use.
Catalog No. 50-002-15802