Learn More
Medchemexpress LLC ML162 1g
SDP
Supplier:  Medchemexpress LLC HY1000021G
Encompass_Preferred
ML162 is a covalent glutathione peroxidase 4 (GPX4) inhibitor ML162 has a selective lethal effect on mutant RAS oncogene-expressing cell lines1]2]
Catalog No. 50-001-68924