Learn More
Medchemexpress LLC HY-100002 5mg ,ML162 CAS:1035072-16-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1000025MG
Encompass_Preferred
HY-100002 5mg ML162 CAS: 1035072-16-2 ML162 is a covalent glutathione peroxidase 4 (GPX4) inhibitor. ML162 has a selective lethal effect on mutant RAS oncogene-expressing cell lines Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-8948