Learn More
Medchemexpress LLC ML179 (SR-1309) | 1883548-87-5 | 99.7% | 422.44 | 50 MG

Supplier: Medchemexpress LLC HY11032910MG
ML179 (SR-1309) is an inverse LRH1/NR5A2 (Liver receptor homologue-1) agonist with an IC50 of 320 nM. It demonstrates anti-proliferation activity in MDA-MB-231 cells and has potential for research into ER-negative breast cancer. This product is for research use only.
- Inverse LRH1/NR5A2 agonist
- IC50 of 320 nM
- Demonstrates anti-proliferation activity in MDA-MB-231 cells
- Potential for research in ER-negative breast cancer
- Powder storage: -20°C for 3 years, 4°C for 2 years
- Solvent storage: -80°C for 6 months, -20°C for 1 month
- Soluble in DMSO (50 mg/mL)
- Soluble in 10% DMSO and 90% corn oil for in vivo studies (≥ 2.5 mg/mL)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.