Learn More
AXON MEDCHEM LLC ML2-SA1, EVP-22 10MG
SDP
Supplier:  AXON MEDCHEM LLC 298010MG
Encompass
ML2-SA1 is a potent, selective and efficacious activator of TRPML2 with EC50 values of 1.24 μM and 2.38 μM for human and mouse TRPML2, respectively. ML2-SA1 shows high selectivity over h/mTRPML1 and h/mTRPML3 in both calcium imaging and endolysosomal patch-clamp experiments and it does not activate TPC1 nor TPC2.MF C14H13Cl2NOMW 282.17 Purity:100% Soluble in DMSO
Catalog No. NC1899716